Gene Transcripts

Gene Symbol
P2rx5
Entrez Gene ID
113995
Gene Name
purinergic receptor P2X, ligand-gated ion channel, 5
Gene Aliases
P2x5
Chromosome Location
Chr. 10: 59725435 - 59737126 on Build Rnor_6.0
UniGene ID
Rn.10257
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080780.2 NP_542958.2 5 702
XM_006246566.2 XP_006246628.1 4 710
GenBank mRNA X92069.1 - 5 702
X97328.1 - 5 568

View Full Product Details