Gene Transcripts

Gene Symbol
Cntn1
Entrez Gene ID
117258
Gene Name
contactin 1
Gene Aliases
F3
Chromosome Location
Chr. 7: 133400485 - 133588314 on Build Rnor_6.0
UniGene ID
Rn.143291
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057118.2 NP_476459.1 7 744
XM_006242177.2 XP_006242239.1 7 759
XM_008765659.1 XP_008763881.1 6 685
GenBank mRNA D38492.1 - 7 748

View Full Product Details