Gene Transcripts

Gene Symbol
Daxx
Entrez Gene ID
140926
Gene Name
death-domain associated protein
Gene Aliases
Chromosome Location
Chr. 20: 5480103 - 5485957 on Build Rnor_6.0
UniGene ID
Rn.870
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB064671.1 - 2 718

View Full Product Details