Gene Transcripts

Gene Symbol
Erg
Entrez Gene ID
170909
Gene Name
v-ets avian erythroblastosis virus E26 oncogene
Gene Aliases
Chromosome Location
Chr. 11: 35645317 - 35749643 on Build Rnor_6.0
UniGene ID
Rn.47987
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133397.2 NP_596888.1 5, 6 733
GenBank mRNA AB031088.1 - 5, 6 733
BC072519.1 - 5, 6 713 7105647
FQ217228.1 - 5, 6 752

View Full Product Details