Gene Transcripts

Gene Symbol
Cyp3a9
Entrez Gene ID
171352
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 9
Gene Aliases
Cyp3a13, P450olf3
Chromosome Location
Chr. 12: 19074288 - 19114491 on Build Rnor_6.0
UniGene ID
Rn.10489
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147206.2 NP_671739.2 11 1136
GenBank mRNA BC086985.1 - 11 1136
U60085.2 - 11 1112

View Full Product Details