Gene Transcripts

Gene Symbol
Ivd
Entrez Gene ID
24513
Gene Name
isovaleryl-CoA dehydrogenase
Gene Aliases
Chromosome Location
Chr. 3: 110669355 - 110689789 on Build Rnor_6.0
UniGene ID
Rn.147
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012592.2 NP_036724.1 6, 7 706
GenBank mRNA BC088401.1 - 6, 7 706
FQ231497.1 - 5, 6 548
J05031.1 - 6, 7 716

View Full Product Details