Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145097.2 | NP_659565.2 | 4 | 1083 | |
XM_008764741.1 | XP_008762963.1 | 5 | 1311 | ||
GenBank mRNA | BC088111.1 | - | 4 | 1083 | |
FQ209558.1 | - | 4 | 1100 | ||
FQ218781.1 | - | 3 | 851 | ||
FQ218835.1 | - | 4 | 1103 | ||
FQ219263.1 | - | 3 | 851 | ||
FQ219393.1 | - | 4 | 1152 | ||
FQ219554.1 | - | 3 | 852 | ||
U51017.1 | - | 3 | 1044 |