Gene Transcripts

Gene Symbol
Rxra
Entrez Gene ID
25271
Gene Name
retinoid X receptor alpha
Gene Aliases
Chromosome Location
Chr. 3: 6272560 - 6295354 on Build Rnor_6.0
UniGene ID
Rn.108206
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012805.2 NP_036937.2 18
GenBank mRNA AM295012.1 - 18
AM295013.1 - 18
AM392401.1 - 18
L06482.1 - 185

View Full Product Details