Gene Transcripts

Gene Symbol
Zfp394
Entrez Gene ID
252860
Gene Name
zinc finger protein 394
Gene Aliases
Rlzfy, Zfp99, Zkscan14, Znf394
Chromosome Location
Chr. 12: 11179323 - 11187362 on Build Rnor_6.0
UniGene ID
Rn.101891
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145724.1 NP_663776.1 3 1682
GenBank mRNA AF052042.1 - 3 1682
BC100140.1 - 3 1676 7442414

View Full Product Details