Gene Transcripts

Gene Symbol
Pygb
Entrez Gene ID
25739
Gene Name
phosphorylase, glycogen; brain
Gene Aliases
GLYPHOA
Chromosome Location
Chr. 3: 146582708 - 146629504 on Build Rnor_6.0
UniGene ID
Rn.1518
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013188.1 NP_037320.1 11, 12 1501
GenBank mRNA BC166475.1 - 1457
L10668.1 - 11, 12 1401

View Full Product Details