Gene Transcripts

Gene Symbol
Lzts1
Entrez Gene ID
266711
Gene Name
leucine zipper, putative tumor suppressor 1
Gene Aliases
Psdzip70
Chromosome Location
Chr. 16: 22250500 - 22306667 on Build Rnor_6.0
UniGene ID
Rn.3213
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153470.1 NP_703200.1 3 1264
GenBank mRNA AB075607.1 - 3 1264

View Full Product Details