Gene Transcripts

Gene Symbol
Vezf1
Entrez Gene ID
287615
Gene Name
vascular endothelial zinc finger 1
Gene Aliases
Chromosome Location
Chr. 10: 75365811 - 75382012 on Build Rnor_6.0
UniGene ID
Rn.198892
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768166.1 XP_008766388.1 3 865
XM_008768167.1 XP_008766389.1 3 865
GenBank mRNA FQ108271.1 - 2 185

View Full Product Details