Gene Transcripts

Gene Symbol
Chaf1b
Entrez Gene ID
288242
Gene Name
chromatin assembly factor 1, subunit B (p60)
Gene Aliases
Chromosome Location
Chr. 11: 34101255 - 34121371 on Build Rnor_6.0
UniGene ID
Rn.159953
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024741.1 NP_001019912.1 14 1762
GenBank mRNA BC097460.1 - 14 1762

View Full Product Details