Gene Transcripts

Gene Symbol
Agpat6
Entrez Gene ID
290843
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 6
Gene Aliases
RGD1310520
Chromosome Location
Chr. 16: 73615949 - 73649769 on Build Rnor_6.0
UniGene ID
Rn.6835
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047849.1 NP_001041314.1 2 1034
GenBank mRNA BC161809.1 - 1 320
DQ480750.1 - 2 1034

View Full Product Details