Gene Transcripts

Gene Symbol
Slc25a2
Entrez Gene ID
291640
Gene Name
solute carrier family 25 (mitochondrial carrier, ornithine transporter) member 2
Gene Aliases
Chromosome Location
Chr. 18: 30774205 - 30775401 on Build Rnor_6.0
UniGene ID
Rn.218546
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106152.1 NP_001099622.1 1 668

View Full Product Details