Gene Transcripts

Gene Symbol
Arv1
Entrez Gene ID
292097
Gene Name
ARV1 homolog (S. cerevisiae)
Gene Aliases
RGD1310264
Chromosome Location
Chr. 19: 57484720 - 57496539 on Build Rnor_6.0
UniGene ID
Rn.13417
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106197.1 NP_001099667.1 3 458
XM_006255806.2 XP_006255868.1 3 416
XM_008772606.1 XP_008770828.1 3 416

View Full Product Details