Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031569.3 | NP_113757.1 | 3 | 806 | |
XM_006235736.2 | XP_006235798.1 | 4 | 896 | ||
XM_006235738.2 | XP_006235800.1 | 3 | 654 | ||
XM_006235739.2 | XP_006235801.1 | 5 | 930 | ||
XM_008762474.1 | XP_008760696.1 | 2 | 416 | ||
GenBank mRNA | AF115267.1 | - | 3 | 627 | |
AY152731.1 | - | 2 | 565 | ||
D16438.1 | - | 3 | 617 | ||
L28144.1 | - | 3 | 737 | ||
L29419.1 | - | 2 | 700 | ||
L33916.1 | - | 3 | 671 | ||
U01913.1 | - | 3 | 746 | ||
U05239.1 | - | 3 | 748 | ||
U07871.1 | - | 2 | 596 |