Gene Transcripts

Gene Symbol
Ms4a14
Entrez Gene ID
293743
Gene Name
membrane-spanning 4-domains, subfamily A, member 14
Gene Aliases
Sp3111
Chromosome Location
Chr. 1: 227471441 - 227486953 on Build Rnor_6.0
UniGene ID
Rn.92466
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181082.2 NP_851596.1 6 839
GenBank mRNA AY258290.1 - 6 839

View Full Product Details