Gene Transcripts

Gene Symbol
Taar2
Entrez Gene ID
294121
Gene Name
trace amine-associated receptor 2
Gene Aliases
Gpr58
Chromosome Location
Chr. 1: 22578025 - 22581994 on Build Rnor_6.0
UniGene ID
Rn.162403
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008512.1 NP_001008512.1 2 251
GenBank mRNA AY702316.1 - 2 251

View Full Product Details