Gene Transcripts

Gene Symbol
Olr411
Entrez Gene ID
296675
Gene Name
olfactory receptor 411
Gene Aliases
Chromosome Location
Chr. 3: 20852767 - 20853708 on Build Rnor_6.0
UniGene ID
Rn.143028
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000385.1 NP_001000385.1 1 484

View Full Product Details