Gene Transcripts

Gene Symbol
Evpl
Entrez Gene ID
303687
Gene Name
envoplakin
Gene Aliases
Chromosome Location
Chr. 10: 105100538 - 105116802 on Build Rnor_6.0
UniGene ID
Rn.229267
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107066.1 NP_001100536.1 22 4518

View Full Product Details