Gene Transcripts

Gene Symbol
Etv5
Entrez Gene ID
303828
Gene Name
ets variant 5
Gene Aliases
Chromosome Location
Chr. 11: 82194657 - 82252145 on Build Rnor_6.0
UniGene ID
Rn.18397
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107082.1 NP_001100552.1 7 769
XM_008768798.1 XP_008767020.1 7 782

View Full Product Details