Gene Transcripts

Gene Symbol
Ikzf4
Entrez Gene ID
100361923
Gene Name
IKAROS family zinc finger 4
Gene Aliases
Chromosome Location
Chr. 7: 3061424 - 3079613 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006240732.2 XP_006240794.1 7 1122
XM_008765052.1 XP_008763274.1 6 1007

View Full Product Details