Gene Transcripts

Gene Symbol
Adamts3
Entrez Gene ID
305253
Gene Name
ADAM metallopeptidase with thrombospondin type 1, motif 3
Gene Aliases
Chromosome Location
Chr. 14: 19866408 - 20072886 on Build Rnor_6.0
UniGene ID
Rn.218557
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107212.1 NP_001100682.1 21 3429
XM_006250773.2 XP_006250835.1 22 3637
XM_006250774.2 XP_006250836.1 22 3265

View Full Product Details