Gene Transcripts

Gene Symbol
Zmym5
Entrez Gene ID
305911
Gene Name
zinc finger, MYM-type 5
Gene Aliases
RGD1306353
Chromosome Location
Chr. 15: 37039481 - 37061898 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002728259.4 XP_002728305.2 7 1458
XM_006252082.2 XP_006252144.1 6 1362
XM_006252083.2 XP_006252145.1 6 1298

View Full Product Details