Gene Transcripts

Gene Symbol
Pgbd1
Entrez Gene ID
103694097
Gene Name
piggyBac transposable element derived 1
Gene Aliases
Chromosome Location
Chr. 17: 45223846 - 45227812 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771776.1 XP_008769998.1 2 2193

View Full Product Details