Gene Transcripts

Gene Symbol
Me2
Entrez Gene ID
307270
Gene Name
malic enzyme 2, NAD(+)-dependent, mitochondrial
Gene Aliases
Chromosome Location
Chr. 18: 69738686 - 69784099 on Build Rnor_6.0
UniGene ID
Rn.16233
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107376.2 NP_001100846.2 10 1140
XM_006254923.2 XP_006254985.1 10 4287
GenBank mRNA BF522605.1 - 2 133

View Full Product Details