Gene Transcripts

Gene Symbol
Zfp35
Entrez Gene ID
307547
Gene Name
zinc finger protein 35
Gene Aliases
Zfp239
Chromosome Location
Chr. 18: 15883182 - 15898803 on Build Rnor_6.0
UniGene ID
Rn.137400
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013141.1 NP_001013159.1 3 483
GenBank mRNA BC089850.1 - 3 483

View Full Product Details