Gene Transcripts

Gene Symbol
Arsa
Entrez Gene ID
315222
Gene Name
arylsulfatase A
Gene Aliases
Chromosome Location
Chr. 7: 130448291 - 130450817 on Build Rnor_6.0
UniGene ID
Rn.23323
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034933.2 NP_001030105.2 8 1309
GenBank mRNA BC105852.1 - 8 1660
FQ212178.1 - 2271

View Full Product Details