Gene Transcripts

Gene Symbol
Ttbk1
Entrez Gene ID
316229
Gene Name
tau tubulin kinase 1
Gene Aliases
RGD1559518
Chromosome Location
Chr. 9: 16862305 - 16899399 on Build Rnor_6.0
UniGene ID
Rn.234117
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271230.1 NP_001258159.1 8 798

View Full Product Details