Gene Transcripts

Gene Symbol
Usp34
Entrez Gene ID
360990
Gene Name
ubiquitin specific peptidase 34
Gene Aliases
Chromosome Location
Chr. 14: 108060653 - 108264706 on Build Rnor_6.0
UniGene ID
Rn.16057
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271196.1 NP_001258125.1 24 3534

View Full Product Details