Gene Transcripts

Gene Symbol
Cenpn
Entrez Gene ID
361416
Gene Name
centromere protein N
Gene Aliases
Da2-27, Da2-4, RGD1310953
Chromosome Location
Chr. 19: 49457462 - 49479996 on Build Rnor_6.0
UniGene ID
Rn.65502
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008366.1 NP_001008367.1 7 703
XM_006255686.2 XP_006255748.1 7 817
XM_006255687.2 XP_006255749.1 7 735
XM_006255689.2 XP_006255751.1 8 1039
XM_006255690.2 XP_006255752.1 8 825
XM_006255691.2 XP_006255753.1 6 759
XM_006255692.2 XP_006255754.1 6 639
GenBank mRNA AY325257.1 - 825
AY325261.1 - 825
BC085839.1 - 7 703 7135799

View Full Product Details