Gene Transcripts

Gene Symbol
Cdh15
Entrez Gene ID
361432
Gene Name
cadherin 15
Gene Aliases
Chromosome Location
Chr. 19: 55669661 - 55689986 on Build Rnor_6.0
UniGene ID
Rn.18406
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207613.1 NP_997496.1 8 1140
GenBank mRNA AB176538.1 - 8 1140

View Full Product Details