Gene Transcripts

Gene Symbol
Ybey
Entrez Gene ID
361822
Gene Name
ybeY metallopeptidase
Gene Aliases
RGD1308722
Chromosome Location
Chr. 20: 12917111 - 12926587 on Build Rnor_6.0
UniGene ID
Rn.41811
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108529.1 NP_001101999.1 3 398
XM_006256301.2 XP_006256363.1 3 409

View Full Product Details