Gene Transcripts

Gene Symbol
Traf3ip2
Entrez Gene ID
361857
Gene Name
Traf3 interacting protein 2
Gene Aliases
RGD1304768
Chromosome Location
Chr. 20: 44680435 - 44722874 on Build Rnor_6.0
UniGene ID
Rn.9031
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044248.1 NP_001037713.1 2 242
XM_006256532.2 XP_006256594.1 2 459
GenBank mRNA BC100099.1 - 2 242

View Full Product Details