Gene Transcripts

Gene Symbol
Bbs5
Entrez Gene ID
362142
Gene Name
Bardet-Biedl syndrome 5
Gene Aliases
Chromosome Location
Chr. 3: 55886695 - 55907717 on Build Rnor_6.0
UniGene ID
Rn.45505
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108583.2 NP_001102053.1 5, 6 423
GenBank mRNA BC167024.1 - 5, 6 423

View Full Product Details