Gene Transcripts

Gene Symbol
Elp6
Entrez Gene ID
363150
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
RGD1305478, Tmem103
Chromosome Location
Chr. 8: 118525770 - 118541074 on Build Rnor_6.0
UniGene ID
Rn.36164
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108782.2 NP_001102252.1 3 209
GenBank mRNA BC166773.1 - 3 209

View Full Product Details