Gene Transcripts

Gene Symbol
Nabp1
Entrez Gene ID
363227
Gene Name
nucleic acid binding protein 1
Gene Aliases
Obfc2a, RGD1306658
Chromosome Location
Chr. 9: 55050194 - 55057659 on Build Rnor_6.0
UniGene ID
Rn.21433
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014216.1 NP_001014238.1 2, 3 427
GenBank mRNA BC089852.1 - 2, 3 427

View Full Product Details