Gene Transcripts

Gene Symbol
RGD1562673
Entrez Gene ID
363551
Gene Name
similar to Prostatic spermine-binding protein precursor (SBP)
Gene Aliases
Chromosome Location
Chr. 10: 13320716 - 13325129 on Build Rnor_6.0
UniGene ID
Rn.111120
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108823.1 NP_001102293.1 2 191
XM_008767589.1 XP_008765811.1 3 1124
GenBank mRNA BC167085.1 - 531

View Full Product Details