Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109006.1 | NP_001102476.1 | 12 | 1682 | |
XM_006245260.2 | XP_006245322.1 | 13 | 1968 | ||
XM_008767280.1 | XP_008765502.1 | 12 | 1861 | ||
XM_008767281.1 | XP_008765503.1 | 13 | 1689 | ||
XM_008767282.1 | XP_008765504.1 | 12 | 1455 |