Gene Transcripts

Gene Symbol
Olr1631
Entrez Gene ID
405126
Gene Name
olfactory receptor 1631
Gene Aliases
Chromosome Location
Chr. 15: 27484665 - 27485639 on Build Rnor_6.0
UniGene ID
Rn.142441
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000837.1 NP_001000837.1 1 610

View Full Product Details