Gene Transcripts

Gene Symbol
Tgfb1
Entrez Gene ID
59086
Gene Name
transforming growth factor, beta 1
Gene Aliases
Tgfb
Chromosome Location
Chr. 1: 82480875 - 82497196 on Build Rnor_6.0
UniGene ID
Rn.40136
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021578.2 NP_067589.1 1 303
GenBank mRNA AY550025.1 - 1 158
BC076380.1 - 1 303 7107411
X52498.1 - 1 570

View Full Product Details