Gene Transcripts

Gene Symbol
Npffr2
Entrez Gene ID
78964
Gene Name
neuropeptide FF receptor 2
Gene Aliases
Gpr74, Npff2, Npgpr
Chromosome Location
Chr. 14: 20148515 - 20193577 on Build Rnor_6.0
UniGene ID
Rn.64524
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023980.1 NP_076470.1 1 231
XM_006250787.2 XP_006250849.1 2 792
GenBank mRNA AF268900.1 - 1 231

View Full Product Details