Gene Transcripts

Gene Symbol
Soat1
Entrez Gene ID
81782
Gene Name
sterol O-acyltransferase 1
Gene Aliases
Acat-1
Chromosome Location
Chr. 13: 74032622 - 74077759 on Build Rnor_6.0
UniGene ID
Rn.59
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031118.1 NP_112380.1 2 131
GenBank mRNA D86373.1 - 2 131

View Full Product Details