Gene Transcripts

Gene Symbol
Cdh1
Entrez Gene ID
83502
Gene Name
cadherin 1
Gene Aliases
Chromosome Location
Chr. 19: 38768467 - 38838395 on Build Rnor_6.0
UniGene ID
Rn.1303
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031334.1 NP_112624.1 15 2544
GenBank mRNA AB017696.1 - 15 2544
AF177680.1 - 1 115
AJ000540.1 - 1 84

View Full Product Details