Gene Transcripts

Gene Symbol
Polr1b
Entrez Gene ID
83582
Gene Name
polymerase (RNA) I polypeptide B
Gene Aliases
RPA2, Rpo1-2
Chromosome Location
Chr. 3: 121632043 - 121656506 on Build Rnor_6.0
UniGene ID
Rn.88734
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031773.1 NP_113961.1 8 1235
GenBank mRNA AF025424.1 - 8 1235

View Full Product Details