Gene Transcripts

Gene Symbol
Kcnj13
Entrez Gene ID
94341
Gene Name
potassium channel, inwardly rectifying subfamily J, member 13
Gene Aliases
Kir7.1
Chromosome Location
Chr. 9: 94486719 - 94495333 on Build Rnor_6.0
UniGene ID
Rn.14516
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053608.2 NP_446060.1 3 843
XM_008767301.1 XP_008765523.1 2 2178
GenBank mRNA AJ006129.1 - 2 726

View Full Product Details