Gene Transcripts

Gene Symbol
Scn2a
Entrez Gene ID
24766
Gene Name
sodium channel, voltage-gated, type II, alpha subunit
Gene Aliases
NachII, Nav1.2, RII/RIIA, RNSCPIIR, SCN, Scn2a1, Scn2a2, ScpII
Chromosome Location
Chr. 3: 51687910 - 51822008 on Build Rnor_6.0
UniGene ID
Rn.89192
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012647.1 NP_036779.1 27 5973
GenBank mRNA M22254.1 - 27 5972
X03639.1 - 27 5973

View Full Product Details