Gene Transcripts

Gene Symbol
Adora2b
Entrez Gene ID
29316
Gene Name
adenosine A2B receptor
Gene Aliases
Chromosome Location
Chr. 10: 48569563 - 48586366 on Build Rnor_6.0
UniGene ID
Rn.89544
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017161.1 NP_058857.1 1, 2 440
GenBank mRNA AF084241.1 - 1, 2 206
BC161854.1 - 1, 2 448 9024632
M91466.1 - 1, 2 440

View Full Product Details