Gene Transcripts

Gene Symbol
Slc12a4
Entrez Gene ID
29501
Gene Name
solute carrier family 12 (potassium/chloride transporter), member 4
Gene Aliases
Kcc1
Chromosome Location
Chr. 19: 37917003 - 37938943 on Build Rnor_6.0
UniGene ID
Rn.32091
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019229.2 NP_062102.1 2 136
XM_006255482.2 XP_006255544.1 2 1540
GenBank mRNA U55815.1 - 2 136

View Full Product Details